Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitamin D binding protein Mouse anti-Human, Clone: 1E4D10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Vitamin D binding protein |
---|---|
Clone | 1E4D10 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6609
![]() ![]() |
Proteintech Group Inc
661751IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.Specifications
Vitamin D binding protein | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DBP, DBP/GC, GC, Gc globulin, VDBP,GC, Vitamin D binding protein | |
GC | |
IgG1 | |
Protein G | |
Antibody |
1E4D10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P02774 | |
2638 | |
Vitamin D binding protein Fusion Protein Ag9803 | |
Primary | |
-20°C | |
GC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title