Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D BP Mouse anti-Human, Clone: 2G2, Novus Biologicals™
SDP

Catalog No. NBP22354101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22354101 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP22354101 Supplier Novus Biologicals Supplier No. NBP2235410.1ML

Mouse Monoclonal Antibody

Vitamin D BP Monoclonal specifically detects Vitamin D BP in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence.

Specifications

Antigen Vitamin D BP
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2G2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunofluorescence 1:10 - 1:50
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias DBP/GC, DBPVDBG, EC 6.3.1.5, Gc-globulin, GRD3, Group-specific component, group-specific component (vitamin D binding protein), hDBP, VDB, VDBP, vitamin D-binding alpha-globulin, vitamin D-binding protein
Gene Symbols GC
Host Species Mouse
Immunogen Gc-globulin isolated from human plasma adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2638
Test Specificity Clone 2G2 is specific for human Gc-globulin.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.