Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D3 Antibody (685503) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB376254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376254 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376254 Supplier Novus Biologicals Supplier No. FAB6566I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vitamin D3 Monoclonal antibody specifically detects Vitamin D3 in Broad Species Reactivity samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Vitamin D3
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 685503
Conjugate Janelia Fluor 549
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen 25-Hydroxyvitamin D3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Broad Species Reactivity
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.