Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitronectin Antibody (019), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB316423 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316423 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316423 Supplier Novus Biologicals Supplier No. NBP290648MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Vitronectin Monoclonal antibody specifically detects Vitronectin in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Vitronectin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 019
Conjugate mFluor Violet 610 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Vitronectin (Accession#: NP_035837.1; Met1-Lys478).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 7448
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.