Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Viva™ HPLC Columns with C18 Column Packing, 3μm Particle Size
SDP

Catalog No. 06739203 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
Flow Rate:
0.07 mL/min
0.31 mL/min
1.5 mL/min
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Flow Rate
06-739-203 150 mm 4.6 mm 1.5 mL/min
06-739-197 50 mm 1 mm 0.07 mL/min
06-739-192 100 mm 4.6 mm 1.5 mL/min
06-739-201 150 mm 2.1 mm 0.31 mL/min
06-739-190 100 mm 2.1 mm 0.31 mL/min
06-739-200 150 mm 1 mm 0.07 mL/min
06-739-196 30 mm 4.6 mm 1.5 mL/min
06-739-199 50 mm 4.6 mm 1.5 mL/min
06-739-198 50 mm 2.1 mm 0.31 mL/min
06-739-194 30 mm 2.1 mm 0.31 mL/min
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 06-739-203 Supplier Restek™ Supplier No. 9514365
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for separating peptides or proteins.

  • Rugged, spherical particles with 300 angstrom pore size
  • High proportion of pore/surface area available to large molecules
  • General-purpose Restek C18 is a conventional monomeric octadecylsilane column
  • Particle Size: 3μm
  • USP Phase Code: L1

Specifications

Diameter (Metric) 4.6 mm
Film Thickness 3 μm
Flow Rate 1.5 mL/min
For Use With Proteins and Other Higher Molecular Weight Compounds
Length (Metric) 150 mm
Particle Shape Spherical
Particle Size 3 μm
pH 2.5 to 10
Pore Size 300 Å
Temperature 80°C
USP Type L1
Stationary Phase C18
Product Line Viva
Packing Material Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel (316)
Technique Reversed Phase HPLC
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.