Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Vivaspin Turbo 15RC
Shop All Sartorius Products
Click to view available options
MWCO:
10 kDa
100 kDa
5 kDa
50 kDa
Quantity:
12 Pk.
48 Pk.
Description
- Fastest sample concentration with maximum target molecule recoveries
- Simplest, pipette-friendly retentate retrieval
- Highest chemical and temperature resistance

Specifications
Specifications
Filter Type | Ultrafiltration |
Format | Spin Column |
For Use With (Equipment) | Centrifuge |
Material | SBC |
Material (Membrane) | Regenerated Cellulose |
MWCO | 10 kDa |
Sample Volume | 4 to 15 mL |
Sterility | Non-sterile |
Quantity | 12 Pk. |
Type | Centrifugal |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction