Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vivid™ NADP+ , 10mM

Catalog No. P2879
Encompass_Preferred
Click to view available options
Quantity:
0.5 mL

Vivid™ NADP+ , 10mM

Vivid™ NADP+, 10 mM, is a Vivid™ CYP450 Screening Kit component consisting of 10 mM NADP+ in 100 mM potassium phosphate, pH 8.0. Vivid™ NADP+, 10 mM, is included in the Vivid™ screening kits, but may also be purchased separately.

More About Vivid™ CYP450 Screening Kits
Vivid™ CYP450 Screening Kits are high-throughput fluorescence-based assays for detection of enzyme-drug interactions and CYP450 inhibition. Vivid™ CYP450 Screening Kits include Vivid™ Substrate, Vivid™ Fluorescent Standard, reaction buffer, Vivid™ Regeneration System, NADP+, and CYP450 BACULOSOMES™ Plus Reagent. Vivid™ Fluorogenic Substrates are blocked dyes that yield minimal fluorescence signal until cleaved or hydroxylated. Oxidation at either of two potential sites releases the highly fluorescent product. CYP450 BACULOSOMES™ Plus Reagents are microsomes prepared from insect cells expressing a human CYP450 isozyme and human cytochrome P450 reductase. CYP450 BACULOSOMES™ Plus Reagents offer a distinct advantage over human liver microsomes in that only one CYP450 isozyme is expressed, thereby preventing metabolism by other CYP450s.

>> View all Vivid™ CYP450 Screening Kits

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Microplate Reader
Product Line VIVID™
Content And Storage 1 tube, Store at -80 °C
Detection Method Fluorescence
Product Type NADP+
Quantity 0.5 mL
Format Kit
Protein Subtype Coenzymes
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.