Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VKORC1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB045132 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045132 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045132 Supplier Novus Biologicals Supplier No. NBP297577AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VKORC1 Polyclonal specifically detects VKORC1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VKORC1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.4.1, EDTP308, FLJ00289, IMAGE3455200, MST576, phylloquinone epoxide reductase, vitamin K dependent clMST134, vitamin K epoxide reductase complex subunit 1, vitamin K epoxide reductase complex, subunit 1, Vitamin K1 2,3-epoxide reductase subunit 1, vitamin K1 epoxide reductase (warfarin-sensitive), VKCFD2, VKORMGC2694
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VKORC1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79001
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.