Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VLDLR Antibody (rVLDLR/1337), FITC, Novus Biologicals™
SDP

Catalog No. NB156529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB156529 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB156529 Supplier Novus Biologicals Supplier No. NBP314200F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

VLDLR Monoclonal antibody specifically detects VLDLR in Human, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen VLDLR
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rVLDLR/1337
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH
Host Species Mouse
Immunogen Recombinant human VLDLR fragment from c-terminal (Uniprot: P98155)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7436
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.