Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VlsE Rabbit anti-Bacteria, Biotin, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP221653B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221653B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221653B Supplier Novus Biologicals Supplier No. NBP221653B

Rabbit Polyclonal Antibody

VlsE Polyclonal specifically detects VlsE in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen VlsE
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation 1 mmol PBS, contains 0.1% glycerol with 0.1% Sodium Azide, 0.01% Thimerosal
Gene Alias Outer surface protein VlsE
Gene Symbols vlsE
Host Species Rabbit
Immunogen MBP recombinant protein corresponding to Borrelia burgdorferi VlsE protein. (Uniprot: G5IXI6)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1194357.0
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.