Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VMA21 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159918 100 μL
NBP15991820 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP159918 Supplier Novus Biologicals Supplier No. NBP159918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VMA21 Polyclonal specifically detects VMA21 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VMA21
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q3ZAQ7
Gene Alias MEAXMGC131652, MGC125514, myopathy with excessive autophagy, Myopathy with excessive autophagy protein, vacuolar ATPase assembly integral membrane protein VMA21, VMA21 vacuolar H+-ATPase homolog (S. cerevisiae), XMEAMGC125516
Gene Symbols VMA21
Host Species Rabbit
Immunogen Synthetic peptides corresponding to LOC203547(hypothetical protein LOC203547) The peptide sequence was selected from the N terminal of LOC203547. Peptide sequence MERPDKAALNALQPPEFRNESSLASTLKTLLFFTALMITVPIGLYFTTKS.
Molecular Weight of Antigen 11 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203547
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Bovine: 91%; Canine: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.