Learn More
Description
Specifications
Specifications
| Antigen | VMA21 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q3ZAQ7 |
| Gene Alias | MEAXMGC131652, MGC125514, myopathy with excessive autophagy, Myopathy with excessive autophagy protein, vacuolar ATPase assembly integral membrane protein VMA21, VMA21 vacuolar H+-ATPase homolog (S. cerevisiae), XMEAMGC125516 |
| Gene Symbols | VMA21 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
