Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VMAT2 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB362385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362385 Supplier Novus Biologicals Supplier No. NB11068123PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VMAT2 Polyclonal antibody specifically detects VMAT2 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)

Specifications

Antigen VMAT2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2
Host Species Rabbit
Immunogen Reacts with an internal sequence of the human, mouse and rat VMAT2 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6571
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.