Learn More
Description
Specifications
Specifications
| Antigen | VN1R1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | G-protein coupled receptor GPCR24, V1RL1hGPCR24, V1R-like 1, V1r-like receptor 1, V3r-related gene protein, VNR19I1, vomeronasal 1 receptor 1, Vomeronasal olfactory receptor chromosome 19 subtype I member 1, vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1, vomeronasal type-1 receptor 1, ZVNH1, ZVNR1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human VN1R1 (NP_065684). Peptide sequence VQHNHSNRLSCRPSQEARATHTIMVLVSSFFVFYSVHSFLTIWTTVVANP |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
