Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VN1R5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152191 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152191 Supplier Novus Biologicals Supplier No. NBP152191
Add to Cart
Add to Cart

Goat Polyclonal Antibody

VN1R5 Polyclonal specifically detects VN1R5 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen VN1R5
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_776257.1
Gene Alias G-protein coupled receptor GPCR26, hGPCR26, V1RL5pheromone receptor, V1R-like 5, V1r-like receptor 5, vomeronasal 1 receptor 5, vomeronasal 1 receptor 5 (gene/pseudogene), vomeronasal type-1 receptor 5
Gene Symbols VN1R5
Host Species Goat
Immunogen Peptide with sequence KFNRQCGSTK corresponding to internal region according to NP_776257.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317705
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.