Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VNN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | VNN2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
VNN2 Polyclonal specifically detects VNN2 in Human samples. It is validated for Western Blot.Specifications
VNN2 | |
Polyclonal | |
Rabbit | |
NP_004656 | |
8875 | |
The immunogen for this antibody is VNN2. Peptide sequence FRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.5.1, EC 3.5.1.92, FOAP-4, Glycosylphosphatidyl inositol-anchored protein GPI-80, GPI-80, GPI-80 variant protein 1, GPI-80 variant protein 2, GPI-80 variant protein 3, GPI-80 variant protein 4, pantetheinase, Protein FOAP-4, vanin 2, vanin-2, Vannin 2, vascular non-inflammatory molecule 2 | |
VNN2 | |
IgG | |
57 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title