Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VNN2 Recombinant Rabbit Monoclonal Antibody (018), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA543152
This item is not returnable.
View return policy
Description
Specificity: Human VNN2/Vanin-2
This gene product is a member of the Vanin family of proteins that share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications
VNN2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O95498 | |
VNN2 | |
Recombinant Human VNN2/Vanin-2 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
018 | |
Unconjugated | |
VNN2 | |
FOAP-4; Glycosylphosphatidyl inositol-anchored protein GPI-80; GPI-80; LOW QUALITY PROTEIN: vascular non-inflammatory molecule 2; pantetheinase; Protein FOAP-4; vanin 2; Vanin-2; Vannin 2; vascular non-inflammatory molecule 2; VNN2 | |
Rabbit | |
Protein A | |
RUO | |
8875 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction