Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ VO-OHpic

Click to view available options
Quantity:
10 mg
50 mg
Description
VO-OHpic is a potent inhibitor of PTEN (phosphatase and tensin homolog) (IC50 = 35 nM). Displays selectivity for PTEN over cysteine-based phosphatases such as SopB, MTM, PTPβ and SAC (IC50 values are 588 nM, 4.03, 57.5 and >10 μM respectively). Increases PIP2 and PIP3 levels; also increases Akt translocation in NIH/3T3 fibroblasts. Blocks the immunosuppressant activity of IDO-activated Tregs. Promotes antitumor inflammatory response in combination with immunotherapy and enhances anticancer chemotherapy in tumor-bearing mice.
Specifications
Specifications
Quantity | 50 mg |
Product Type | VO-OHpic |
Purity | >98% |
Molecular Weight (g/mol) | 361.16 |
Inhibitors | VO-OHpic |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction