Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Hippel Lindau Antibody (AT82B10), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP1043A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP1043A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP1043A94 Supplier Novus Biologicals Supplier No. NBP104355AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Von Hippel Lindau Monoclonal specifically detects Von Hippel Lindau in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Von Hippel Lindau
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AT82B10
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor
Gene Symbols VHL
Host Species Mouse
Immunogen Recombinant human His-VHL (1-154 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, HIF Target Genes, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7428.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.