Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Antibody (111), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB099712 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099712 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099712 Supplier Novus Biologicals Supplier No. NBP289724AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.

Specifications

Antigen Von Willebrand Factor
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 111
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Von Willebrand Factor (Met 1-Lys 2813; NP_000543.2) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.