Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Antibody, Novus Biologicals™
SDP

Catalog No. NB60058601M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB60058601M 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB60058601M Supplier Novus Biologicals Supplier No. NB6005860.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

Von Willebrand Factor Polyclonal specifically detects Von Willebrand Factor in Human, Mouse, Canine, Feline samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dual RNAscope ISH-IHC.

Specifications

Antigen Von Willebrand Factor
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:1000
Formulation 10mM PBS (pH 7.4) with 15mM Sodium Azide
Gene Accession No. P04275
Gene Alias coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Gene Symbols VWF
Host Species Rabbit
Immunogen Purified antisera of the Von Willebrand Factor (factor VIII R:ag) related antigen in plasma.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Test Specificity The antibody specifically reacts with the cytoplasm of human endothelial cells from normal and neoplastic blood and from lymphatic vessels. It also reacts with human endocardium, platelets and megakaryocytes. It may be used for immunohistological studies of the vascular structures in frozen tissue preparations and enzymatically pre-digested, formalin, methacarn, Bouin's or ethanol-fixed and paraffin-embedded tissue sections.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.