Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Antibody (rVWF/1465) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119688 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119688 Supplier Novus Biologicals Supplier No. NBP308962
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Von Willebrand Factor
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rVWF/1465
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 ug/mL, Immunoprecipitation : 0.5-1 ug/500 ug protein lysate, Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Host Species Mouse
Immunogen Recombinant fragment of human Von Willebrand Factor protein (aa1815-1939) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.