Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VP1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB210109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB210109 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB210109 Supplier Novus Biologicals Supplier No. NB10093575AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VP1 Polyclonal antibody specifically detects VP1 in Adeno-associated Virus (AAV) samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen VP1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias AAV, AAV2, Adeno-Associated Virus, Adeno-Associated Virus (AAV)
Host Species Rabbit
Immunogen Synthetic peptide within residues 70-90 of major coat protein VP3 [AAV 2], residues 270-290 of capsid protein VP1 [AAV 6], and residues 270-290 of capsid [AAV 3].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Gene Therapy, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Adeno-associated Virus (AAV)
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.