Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VPARP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120485 Supplier Invitrogen™ Supplier No. PA5120485
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, Mouse heart. The target is usually found in the following locations: Cytoplasm, Nucleus, cytoskeleton, spindle. Immunogen sequence: MVMGIFANCI FCLKVKYLPQ QQKKKLQTDI KENGGKFSFS LNPQCTHIIL DNADVLSQYQ LNSIQKNHVH IANPDFIWKS IREKRLLDVK NYDPYKPLDI TPPPDQKASS SEVKTEGLCP DSATEEEDTV ELTEFGMQNV EIPHLPQDFE VAKYNTLEKV GMEGGQEAVV VELQCSRDSR DCPFLISSHF LLDDGMETRR QFAIKKTSED ASEYFENYIE ELKKQGFLLR EHFTPEATQL ASEQLQALLL EEVMNSSTLS QEVSDLVEMI WAEALGHLEH MLLKPVNRIS LNDVSKAEGI.

This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPARP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PARP4
Gene Accession No. Q9UKK3
Gene Alias 193 kDa vault protein; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 1; ADP-ribosyltransferase diphtheria toxin-like 4; ADPRTL1; ARTD4; C030027K23Rik; E230037B21Rik; Gm743; H5 proline-rich; I-alpha-I-related; KIAA0177; p193; Parp4; PARP-4; PARPL; PARP-related; PARP-related/IalphaI-related H5/proline-rich; PH5P; poly (ADP-ribose) polymerase family, member 4; poly (ADP-ribose) polymerase)-like 1; poly [ADP-ribose] polymerase 4; poly(ADP-ribose) polymerase family member 4; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase-like 1; Protein mono-ADP-ribosyltransferase PARP4; Vault poly(ADP-ribose) polymerase; vault protein, 193-kDa; VAULT3; von Willebrand factor A domain containing 5C; VPARP; VWA5C
Gene Symbols PARP4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PARP4 (NP_006428.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143, 328417, 361046
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.