Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VprBP Antibody, Novus Biologicals™
SDP

Catalog No. NBP227203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227203 0.1 mg
NBP227203SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227203 Supplier Novus Biologicals Supplier No. NBP227203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VprBP Polyclonal specifically detects VprBP in Human, Mouse, Rat, Porcine, Chicken, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen VprBP
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml∼, Flow Cytometry 0.2ug/10^6cells
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9Y4B6
Gene Alias DCAF1DDB1 and CUL4 associated factor 1, DDB1- and CUL4-associated factor 1, HIV-1 Vpr-binding protein, KIAA0800protein VPRBP, MGC102804, RIP, Vpr (HIV-1) binding protein, Vpr-binding protein, VprBP, Vpr-interacting protein
Gene Symbols VPRBP
Host Species Rabbit
Immunogen A portion of amino acids 935-985 from human VPRBP was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9730
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, european pole-cat, pig, orangutan, rabbit, white-cheeked gibbon, opossum, wild turkey, african bush elephant, chicken, horse, Chinese hamster, guinea pig, and panda, 94% in cow, and platypus, and 88% homolgous in western clawed frog.There are numerous isoforms including 1505, 1504, 1076 and 783 amino acids.
Target Species Human, Mouse, Rat, Pig, Chicken, Chinese Hamster, Equine, Feline, Guinea Pig, Opossum, Other, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.