Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS11 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB290612 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290612 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290612 Supplier Novus Biologicals Supplier No. NBP204093F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS11 Polyclonal antibody specifically detects VPS11 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen VPS11
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias hVPS11, PEP5, RING finger protein 108, RNF108END1, vacuolar protein sorting 11 (yeast homolog), vacuolar protein sorting 11 homolog (S. cerevisiae), vacuolar protein sorting protein 11, vacuolar protein sorting-associated protein 11 homolog
Host Species Rabbit
Immunogen Maps to a region between residue 891 and 941 of human Vacuolar Protein Sorting 11 using the numbering given in entry NP_068375.3 (GeneID 55823).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 55823
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.