Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS16 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB047440 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047440 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047440 Supplier Novus Biologicals Supplier No. NBP297757DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS16 Polyclonal specifically detects VPS16 in Human samples. It is validated for Western Blot.

Specifications

Antigen VPS16
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias hVPS16, vacuolar protein sorting 16 homolog (S. cerevisiae), vacuolar protein sorting protein 16, vacuolar protein sorting-associated protein 16 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VPS16 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.