Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS24 Rabbit anti-Human, Mouse, Rat, Clone: 1L10B7, Novus Biologicals™
SDP

Catalog No. NB168887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168887 100 μg
NB168888 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168887 Supplier Novus Biologicals Supplier No. NBP316902100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VPS24 Monoclonal antibody specifically detects VPS24 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen VPS24
Applications Western Blot
Classification Monoclonal
Clone 1L10B7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CGI-149, CHMP325.1 protein, Chromatin-modifying protein 3, hVps24, member 3,25.1, NEDFFLJ38114, vacuolar protein sorting 24 (yeast), vacuolar protein sorting 24 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 65-220 of human VPS24 (Q9Y3E7).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51652
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.