Learn More
Description
Specifications
Specifications
| Antigen | VPS29 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000 |
| Formulation | Lyophilized from whole antisera with No Preservative |
| Gene Accession No. | Q9QZ88 |
| Gene Alias | DC15, DC7, DKFZp564F0223, EC 3.1.3.3, FLJ20492, hVPS29, PEP11, PEP11 homolog, retromer protein, vacuolar protein sorting 29 (yeast homolog), vacuolar protein sorting 29 (yeast), vacuolar protein sorting 29 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 29, vacuolar sorting protein VPS29/PEP11, Vesicle protein sorting 29, x 007 protein |
| Gene Symbols | VPS29 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
