Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS34 Antibody, Novus Biologicals™
SDP

Catalog No. NB11087320T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11087320T 0.025 mL
NB11087320 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11087320T Supplier Novus Biologicals Supplier No. NB11087320SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

VPS34 Polyclonal specifically detects VPS34 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen VPS34
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100, Knockdown Validated
Formulation PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide
Gene Accession No. Q8NEB9
Gene Alias EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI3K type 3, PI3-kinase type 3, PtdIns-3-kinase type 3, Vps34
Gene Symbols PIK3C3
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human VPS34 protein (within residues 700-850). [Swiss-Prot# Q8NEB9]
Molecular Weight of Antigen 100 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5289
Test Specificity Human, rat, mouse and porcine. Immunogen sequence has 87% homology to Xenopus.
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.