Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS35 Antibody (11H10), Novus Biologicals™
SDP

Catalog No. NB002994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002994 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002994 Supplier Novus Biologicals Supplier No. NBP278827
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VPS35 Monoclonal specifically detects VPS35 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen VPS35
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 11H10
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 1:200
Formulation PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide
Gene Alias DKFZp434P1672, FLJ10752, FLJ13588, FLJ20388, hVPS35, Maternal-embryonic 3, MEM3DKFZp434E1211, vacuolar protein sorting 35 (yeast homolog), vacuolar protein sorting 35 (yeast), vacuolar protein sorting 35 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 35, Vesicle protein sorting 35
Gene Symbols VPS35
Host Species Mouse
Immunogen Full length recombinant human VSP35
Purification Method Protein G purified
Quantity 100 μg
Research Discipline Autophagy, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 55737
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.