Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS35 Antibody, Novus Biologicals™
SDP

Catalog No. NB434203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB434203 25 μg
NB1001397 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB434203 Supplier Novus Biologicals Supplier No. NB100139725UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 22 publications

VPS35 Polyclonal specifically detects VPS35 in Canine, Chicken, Chinese Hamster, Human, Mouse, Rat samples. It is validated for IF/IHC, IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, WB, Western Blot.

Specifications

Antigen VPS35
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.1 μg/mL, Immunohistochemistry 8 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunohistochemistry-Paraffin 8 μg/mL, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_060676.2
Gene Alias DKFZp434P1672, FLJ10752, FLJ13588, FLJ20388, hVPS35, Maternal-embryonic 3, MEM3DKFZp434E1211, vacuolar protein sorting 35 (yeast homolog), vacuolar protein sorting 35 (yeast), vacuolar protein sorting 35 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 35, Vesicle protein sorting 35
Gene Symbols VPS35
Host Species Goat
Immunogen Peptide with sequence C-SPESEGPIYEGLIL corresponding to C-Terminus according to NP_060676.2.
Molecular Weight of Antigen 91 kDa
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55737
Test Specificity Note there is a hypothetical protein called similar to vacuolar protein sorting 35 (XP_040192.1), which is virtually identical.
Target Species Canine, Mouse, Chicken, Chinese Hamster, Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.