Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VPS35 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 22 publications
$231.00 - $562.50
Specifications
Antigen | VPS35 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
VPS35 Polyclonal specifically detects VPS35 in Human, Mouse, Rat, Canine, Chicken, Chinese Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
VPS35 | |
Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
RUO | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DKFZp434P1672, FLJ10752, FLJ13588, FLJ20388, hVPS35, Maternal-embryonic 3, MEM3DKFZp434E1211, vacuolar protein sorting 35 (yeast homolog), vacuolar protein sorting 35 (yeast), vacuolar protein sorting 35 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 35, Vesicle protein sorting 35 | |
VPS35 | |
IgG | |
Affinity Purified | |
91 kDa |
0.5 mg/mL | |
Polyclonal | |
Goat | |
Human, Mouse, Chinese Hamster | |
NP_060676.2 | |
55737 | |
Peptide with sequence C-SPESEGPIYEGLIL corresponding to C-Terminus according to NP_060676.2. | |
Primary | |
Note there is a hypothetical protein called similar to vacuolar protein sorting 35 (XP_040192.1), which is virtually identical. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title