Learn More
Description
Specifications
Specifications
| Antigen | VPS35 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.93mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7), 20% Glycerol with 0.025% Proclin 300 |
| Gene Accession No. | Q96QK1 |
| Gene Alias | DKFZp434P1672, FLJ10752, FLJ13588, FLJ20388, hVPS35, Maternal-embryonic 3, MEM3DKFZp434E1211, vacuolar protein sorting 35 (yeast homolog), vacuolar protein sorting 35 (yeast), vacuolar protein sorting 35 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 35, Vesicle protein sorting 35 |
| Gene Symbols | VPS35 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
