Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS37B Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB135007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135007 Supplier Novus Biologicals Supplier No. NBP306284V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS37B Polyclonal specifically detects VPS37B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VPS37B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ESCRT-I complex subunit VPS37B, FLJ12750, hVps37B, vacuolar protein sorting 37 homolog B (S. cerevisiae), vacuolar protein sorting 37B, vacuolar protein sorting 37B (yeast), vacuolar protein sorting-associated protein 37B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VPS37B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79720
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.