Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS41 Rabbit anti-Human, Mouse, Rat, Clone: S08-1J7, Novus Biologicals™
SDP

Catalog No. NB165424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165424 100 μg
NB165425 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165424 Supplier Novus Biologicals Supplier No. NBP315109100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VPS41 Monoclonal antibody specifically detects VPS41 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen VPS41
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S08-1J7
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias HVPS41, hVps41p, HVSP41, S53, vacuolar assembly protein 41, vacuolar protein sorting 41 (yeast homolog), vacuolar protein sorting 41 (yeast), vacuolar protein sorting 41 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 41 homolog
Host Species Rabbit
Immunogen A synthetic peptide of human VPS41 (Uniprot#: P49754)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cellular Markers, Microautophagy
Primary or Secondary Primary
Gene ID (Entrez) 27072
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.