Learn More
Description
Specifications
Specifications
| Antigen | VPS4A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| Gene Accession No. | Q9UN37 |
| Gene Alias | FLJ22197, hVPS4, Protein SKD2, SKD1, SKD1A, SKD2, vacuolar protein sorting 4 homolog A (S. cerevisiae), vacuolar protein sorting 4A (yeast homolog), vacuolar protein sorting factor 4A, vacuolar protein sorting-associated protein 4A, vacuolar sorting protein 4, VPS4-1SKD1-homolog, VPS4vacuolar protein sorting 4A (yeast) |
| Gene Symbols | VPS4A |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
