Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS4B Antibody, Novus Biologicals™
SDP

Catalog No. NBP119118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119118 Supplier Novus Biologicals Supplier No. NBP119118

Rabbit Polyclonal Antibody

VPS4B Polyclonal specifically detects VPS4B in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen VPS4B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_004860.2
Gene Alias Cell migration-inducing gene 1 protein, Protein SKD1, SKD1B, SKD1cell migration-inducing 1, Suppressor of K(+) transport growth defect 1, suppressor of K+ transport defect 1, vacuolar protein sorting 4 homolog B (S. cerevisiae), vacuolar protein sorting 4B, vacuolar protein sorting 4B (yeast), vacuolar protein sorting-associated protein 4B, VPS42, VPS4-2
Gene Symbols VPS4B
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human vacuolar protein sorting 4 homolog B using the numbering given in entry NP_004860.2 (GeneID 9525).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 9525
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.