Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$518.60

Specifications

Antigen VPS4B
Concentration 0.23 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-173-1714
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
176731AP150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

Class E vacuolar protein sorting (vps) proteins are necessary for appropriate sorting of receptors in the yeast endocytic pathway. The yeast Vps4p is a member of the AAA protein family (ATPases associated with diverse cellular activities) and plays an important role in transporting proteins out of a prevacuolar endosomal compartment. In human, two non-allelic orthologous proteins (VPS4A and VPS4B) of yeast Vps4p are known and can form heteromeric complexes with each other. Both VPS4 (also known as SKD1 in mouse) proteins are class E VPSs and are involved in intracellular protein trafficking, similar to Vps4p in yeast. A human CHMP1 protein, which is implicated in multivesicular body formation, physically interacts with VPS4. HIV-1 uses cellular machinery to bud from infected cells and requires VPS4 and TSG101/VPS23 for this budding process. Domit negative mutant of VPS4 inhibit vacuolar protein sorting and also arrest HIV-1 and MLV budding. Thus, retroviruses normally use the VPS pathway to form multivesicular bodies during the budding process.
Specifications

Specifications

VPS4B
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O75351, P46467
20479, 360834, 9525
VPS4B Fusion Protein Ag11610
Primary
-20°C
VPS4B
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
MIG1, Protein SKD1, SKD1, VPS4 2, VPS42, VPS4B
Vps4b
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.