Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS50 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178217 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178217 Supplier Novus Biologicals Supplier No. NBP178217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS50 Polyclonal specifically detects VPS50 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen FLJ20097
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_060137.2
Gene Alias coiled-coil domain containing 132, coiled-coil domain-containing protein 132, DKFZp313I2429, FLJ20097, FLJ23581, KIAA1861MGC176659
Gene Symbols CCDC132
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 914 and 964 of human Coiled-Coil Domain Containing 132 using the numbering given in entry NP_060137.2 (GeneID 55610).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55610
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.