Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS52 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB050366 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050366 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050366 Supplier Novus Biologicals Supplier No. NBP297979F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS52 Polyclonal specifically detects VPS52 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VPS52
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ARE1, dJ1033B10.5, DKFZp547I194, SAC2, SAC2 suppressor of actin mutations 2-like (yeast), SAC2 suppressor of actin mutations 2-like protein, SACM2LRP5-1033B10, vacuolar protein sorting 52 (yeast), vacuolar protein sorting 52 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 52 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VPS52 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 6293
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.