Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VPS53 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB047499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047499 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047499 Supplier Novus Biologicals Supplier No. NBP297767AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VPS53 Polyclonal specifically detects VPS53 in Human samples. It is validated for Western Blot.

Specifications

Antigen VPS53
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ10979, FLJ41112, FLJ61757, HCCS1, hVps53L, MGC39512, pp13624, vacuolar protein sorting 53 (yeast), vacuolar protein sorting 53 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 53 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VPS53 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55275
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.