Learn More
Description
Specifications
Specifications
| Antigen | VRK1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant VRK1 (Q99986; Met1-Lys396). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
