Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VRK1 Antibody (OTI2C9), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274855B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274855B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274855B Supplier Novus Biologicals Supplier No. NBP274855B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VRK1 Monoclonal specifically detects VRK1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen VRK1
Applications Western Blot
Classification Monoclonal
Clone OTI2C9
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1
Gene Symbols VRK1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-230 of human VRK1 (NP_003375) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7443.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.