Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VRK2 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294871 Supplier Novus Biologicals Supplier No. NBP325746

Rabbit Polyclonal Antibody

VRK2 Polyclonal antibody specifically detects VRK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen VRK2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH7), 20% Glycerol
Gene Alias EC 2.7.11.1, serine/threonine-protein kinase VRK2, vaccinia related kinase 2, vaccinia virus B1R-related kinase 2, Vaccinia-related kinase 2, vaccinia-related kinase-2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VRK2. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 7444
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.