Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG1 Antibody (1314D), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB369012 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369012 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369012 Supplier Novus Biologicals Supplier No. FAB48181E
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VSIG1 Monoclonal specifically detects VSIG1 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen VSIG1
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1314D
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Cell surface A33 antigen, dJ889N15.1, Glycoprotein A34, GPA34, MGC44287,1700062D20Rik, V-set and immunoglobulin domain containing 1, V-set and immunoglobulin domain-containing protein 1
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human VSIG-1, Val22-Gly234, Accession # Q86XK7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.