Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG2 Antibody (541522), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB312131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312131 Supplier Novus Biologicals Supplier No. FAB51771PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSIG2 Monoclonal antibody specifically detects VSIG2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen VSIG2
Applications Flow Cytometry
Classification Monoclonal
Clone 541522
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias 2210413P10Rik, Cortical thymocyte-like protein, CTHCTXLcortical thymocyte receptor (X. laevis CTX) like, CT-like protein, V-set and immunoglobulin domain containing 2, V-set and immunoglobulin domain-containing protein 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VSIG2, Val24-Ala243, Accession # Q961Q7-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23584
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.