Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG2 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
SDP

Catalog No. p-200011066
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1780 30 μL
50-167-3204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1780 Supplier OriGene Supplier No. TA502124S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSIG2 Monoclonal antibody specifically detects VSIG2 in Human, Rat samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen VSIG2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1H2
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ANXA3
Gene Accession No. P12429, P14669
Gene Alias 1190004B15Rik; 2210413P10Rik; AU040484; cortical thymocyte receptor (X. laevis CTX) like; cortical thymocyte-like protein; CTH; CT-like protein; Ctm; CTX; CTXL; RGD1561274; UNQ2770/PRO7154; V-set and immunoglobulin domain containing 2; V-set and immunoglobulin domain-containing protein 2; VSIG2
Host Species Mouse
Immunogen Full length human recombinant protein of human ANXA3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25291, 306
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.