Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG3/IGSF11 Antibody (973422), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB310521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310521 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310521 Supplier Novus Biologicals Supplier No. FAB92293PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VSIG3/IGSF11 Monoclonal antibody specifically detects VSIG3/IGSF11 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen VSIG3/IGSF11
Applications Flow Cytometry
Classification Monoclonal
Clone 973422
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human VSIG3, Met1-Gly245, Accession # Q5DX21
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152404
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.