Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSIG4 Antibody (202), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB334718 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334718 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334718 Supplier Novus Biologicals Supplier No. NBP290066MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen VSIG4
Applications ELISA
Classification Monoclonal
Clone 202
Conjugate mFluor Violet 500 SE
Dilution ELISA, Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human VSIG4 (Uniprot#: Q9Y279-1, Met1-Pro283)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11326
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.